Silver iodide, 99.9% (metals basis)
Silver iodide, 99.9% (metals basis)
Silver iodide, 99.9% (metals basis)
Silver iodide, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver iodide, 99.9% (metals basis)

Silver iodide, AgI, CAS Number-7783-96-2, colloidal silver iodide, silver iodide, ccris 8490, neosilvol, silver 1+ iodide, iodanylsilver, neosiluol, silver monoiodide, silver iodide metals basis, 100g, 99.9% (metals basis), 5.67 g/mL, Hazard Class: 9; Packaging Group: III, 232-038-0, 234.77 | CAS: 7783-96-2 | AgI | 234.773 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011419.14
Price (EUR)
219,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
219,00
Each
Add to cart
Chemical Identifiers
CAS7783-96-2
IUPAC Namesilver(1+) iodide
Molecular FormulaAgI
InChI KeyMSFPLIAKTHOCQP-UHFFFAOYSA-M
SMILES[Ag+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Used in medicine for its caustic, astringent and antiseptic effects. Used in photography, and in the estimation of silver. Through heterogeneous nucleation, it is used in cloud seeding, relying on its beta-AgI crystal structure being similar to that of Ice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only