Iridium foil, 0.25mm (0.01in) thick, hard, 99.8% (metals basis)
Iridium foil, 0.25mm (0.01in) thick, hard, 99.8% (metals basis)
Iridium foil, 0.25mm (0.01in) thick, hard, 99.8% (metals basis)
Iridium foil, 0.25mm (0.01in) thick, hard, 99.8% (metals basis)
Thermo Scientific Chemicals

Iridium foil, 0.25mm (0.01in) thick, hard, 99.8% (metals basis)

Form: approx. 3.5g/25 x 25mm | CAS: 7439-88-5 | Ir | 192.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 011427.FI
also known as 011427-FI
Price (EUR)
-
Quantity:
50 x 50 mm
Request bulk or custom format
Chemical Identifiers
CAS7439-88-5
IUPAC Nameiridium
Molecular FormulaIr
InChI KeyGKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES[Ir]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.25mm thick, hard
Impurity contentList metal impurities (reported)
Total Metal Impurities≤0.2%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only