Iridium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)
Iridium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)
Iridium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)
Iridium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)
Thermo Scientific Chemicals

Iridium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)

CAS: 7439-88-5 | Ir | 192.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 cm
1 cm
Catalog number 011429.BQ
also known as 011429-BQ
Price (USD)/ Each
-
Quantity:
5 cm
Request bulk or custom format
Chemical Identifiers
IUPAC Nameiridium
Molecular FormulaIr
InChI KeyGKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES[Ir]
Molecular Weight (g/mol)192.22
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 1.0mm (0.04in) thick.
Assay from Supplier's CofA≥99.8% (metals basis)
Appearance (Color)Gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only