Thermo Scientific Chemicals

Cobalt rod, 5mm (0.2in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 011445.DO
150 mm, Each
Thermo Scientific Chemicals

Cobalt rod, 5mm (0.2in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 011445.DO
150 mm, Each
Quantity
Catalog number: 011445.DO
also known as 011445-DO
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7440-48-4
IUPAC Name
cobalt
Molecular Formula
Co
InChI Key
GUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES
[Co]
Form
Rod, 5 mm (0.2 in.) dia.
Total Metal Impurities
≤0.005%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Respiratory Sensitization
    Category 1
    Respiratory Sensitization
    Sub-category 1B
    Skin Sensitization
    Category 1
    Germ Cell Mutagenicity
    Category 2
    Carcinogenicity
    Category 1B
    Reproductive Toxicity
    Category 1B
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H317 - May cause an allergic skin reaction

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    H341 - Suspected of causing genetic defects

    H350i - May cause cancer by inhalation

    H360F - May damage fertility

    H412 - Harmful to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P284 - Wear respiratory protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

    Additional EU labelling

    Restricted to professional users