Thermo Scientific Chemicals

Antimony(III) fluoride, 99+%, Thermo Scientific Chemicals

Catalog number: 011492.22
100 g, Each
Thermo Scientific Chemicals

Antimony(III) fluoride, 99+%, Thermo Scientific Chemicals

Catalog number: 011492.22
100 g, Each
Quantity
Catalog number: 011492.22
also known as 011492-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7783-56-4
IUPAC Name
antimony(3+) trifluoride
Molecular Formula
F3Sb
InChI Key
GUNJVIDCYZYFGV-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Sb+3]
Assay from Suppliers CofA
Fluoride: 31.57-32.20%
Form
Powder or granular powder
Assay from Suppliers CofA
≥99.0% (based on fluoride)
Appearance (Color)
White to grey or pale brown

Description

Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of Freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text