Aluminum nitride, N 32.0% min
Aluminum nitride, N 32.0% min
Aluminum nitride, N 32.0% min
Thermo Scientific Chemicals

Aluminum nitride, N 32.0% min

CAS: 24304-00-5 | AlN | 40.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
1 kg
Catalog number 011546.A1
also known as 011546-A1
Price (EUR)
649,00
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
649,00
Each
Chemical Identifiers
CAS24304-00-5
IUPAC Namealumanylidyneamine
Molecular FormulaAlN
InChI KeyPIGFYZPCRLYGLF-UHFFFAOYSA-N
SMILESN#[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)32.0% N min
Aluminum nitride possesses very high thermal conductivity and effectively used in the ceramic industry. It finds application in deep ultraviolet optoelectronics, steel production, dielectric layers in optical storage media, chip carriers and as a crucible to grow crystals of gallium arsenide. It is also used as a radio frequency filter, which finds application in mobile phones. Further, it is used as a circuit carrier in semiconductors. In addition to this, it is used as a heat-sink in light-emitting diode lighting technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only