Sodium fluoride, ACS, 99% min, Thermo Scientific Chemicals
Sodium fluoride, ACS, 99% min, Thermo Scientific Chemicals
Sodium fluoride, ACS, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium fluoride, ACS, 99% min, Thermo Scientific Chemicals

Steel Degreasing Agent, in Tin Electroplating, Flux
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 011561.30
價格 (TWD)
-
Quantity:
250 g
Request bulk or custom format
化學識別
CAS7681-49-4
IUPAC Namesodium fluoride
Molecular FormulaFNa
InChI KeyPUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES[F-].[Na+]
檢視更多
規格Specification Sheet規格表
Assay (Titration)99% NaF min
Free acid (titration)0.03meq/g, max
Free base (titration)0.01meq/g, max
Impurity contentCl 0.005%, max
Impurity contentFe 0.003%, max
檢視更多
It is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only