Thermo Scientific Chemicals

Mangan(II)-chlorid-Tetrahydrat, Thermo Scientific Chemicals

Katalognummer: 011563.36
500 g, Each
Thermo Scientific Chemicals

Mangan(II)-chlorid-Tetrahydrat, Thermo Scientific Chemicals

Katalognummer: 011563.36
500 g, Each
Menge
Katalognummer: 011563.36
auch als 011563-36 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
13446-34-9
IUPAC Name
manganese(2+) tetrahydrate dichloride
Molecular Formula
Cl2H8MnO4
InChI Key
CNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILES
O.O.O.O.[Cl-].[Cl-].[Mn++]
Appearance (Color)
Pink
Form
Crystal
Assay (Titration)
98-101% [U.S. specification]
Comment
Specification differs for U.S. and non-U.S. material where indicated
Insoluble material
0.005% max [U.S. specification]

Beschreibung

Manganese(II) chloride tetrahydrate is used in the tailing of DNA - MnCl2 buffer is recommended for dC and dG tailing and in reverse transcription of RNA with thermostable DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Serious Eye Damage/Eye Irritation
    Category 1
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 - Toxic if swallowed

    H318 - Causes serious eye damage

    H373 - May cause damage to organs through prolonged or repeated exposure

    H411 - Toxic to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician