Thermo Scientific Chemicals

Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 011593.A1
1 kg, Each
Thermo Scientific Chemicals

Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 011593.A1
1 kg, Each
Quantity
Catalog number: 011593.A1
also known as 011593-A1
Price (USD)
Price: 196.00
Online price: 166.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7758-11-4
IUPAC Name
dipotassium hydrogen phosphate
Molecular Formula
HK2O4P
InChI Key
ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES
[K+].[K+].OP([O-])([O-])=O
Heavy Metals
(ICP-OES) 5ppm
Impurity content
Cl 0.003%
Impurity content
Nitrogen compounds (as N) 0.001%
Impurity content
SO₄ 0.005%
Assay (unspecified)
98.0% K₂HPO₄ min

Description

It is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text