Yoduro de potasio, ACS, 99,0 % mín., Thermo Scientific Chemicals
Yoduro de potasio, ACS, 99,0 % mín., Thermo Scientific Chemicals
Yoduro de potasio, ACS, 99,0 % mín., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yoduro de potasio, ACS, 99,0 % mín., Thermo Scientific Chemicals

Para su uso en emulsiones fotográficas, en química analítica, como aditivo de alimentación
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
2 kg
Número de catálogo 011601.36
también denominado 011601-36
Precio (EUR)
176,00
Each
Cantidad:
500 g
Pedido a granel o personalizado
Precio (EUR)
176,00
Each
Identificadores químicos
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Impurity contentIO₃ 3ppm
pHof a 5% solution 6.0-9.2 at 25?
Assay (unspecified)99.0% KI min
Impurity contentCa 0.002%
Impurity contentChloride and bromide (as Cl) 0.01%
Ver más
It is both a source of iodide and a mild reducing agent. It is used as a component in the electrolyte of dye sensitized solar cells. In the field of synthetic organic chemistry, it is used in the preparation of alkyl or aryl iodo compounds. It is employed as a nucleophilic catalyst in the alkylations with alkyl bromide, chloride, tosylate and mesylate. In the presence of acid it forms hydroiodic acid, which is a strong reducing agent. It is useful in producing Lugol’s solution (or Lugol’s iodine) which finds many applications including starch testing, gram stains, and etching of gold surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only