Lithium perchlorate, anhydrous, ACS, 95.0% min
Lithium perchlorate, anhydrous, ACS, 95.0% min
Lithium perchlorate, anhydrous, ACS, 95.0% min
Lithium perchlorate, anhydrous, ACS, 95.0% min
Lithium perchlorate, anhydrous, ACS, 95.0% min
Thermo Scientific Chemicals

Lithium perchlorate, anhydrous, ACS, 95.0% min

As an oxidizing agent | CAS: 7791-03-9 | ClH6LiO7 | 160.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number 011639.09
also known as 011639-09
Price (EUR)
38,20
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)
38,20
Each
Add to cart
Chemical Identifiers
CAS7791-03-9
IUPAC Namelithium(1+) trihydrate perchlorate
Molecular FormulaClH6LiO7
InChI KeyJAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES[Li+].O.O.O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentFe 5ppm
Impurity contentSO₄ 0.001%
pHof a 5% solution 6.0-7.5 at 25?
Heavy Metals(ICP-OES) 5ppm
Assay (unspecified)95.0% LiClO₄ min
View more
Lithium perchlorate is used as an oxygen source for chemical oxygen generators, as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte, and act as chaotropic agent to denature proteins. It is used in the Baylis-Hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only