Thermo Scientific Chemicals

Barium perchlorate trihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011656.30
250 g, Each
Thermo Scientific Chemicals

Barium perchlorate trihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011656.30
250 g, Each
Quantity
Catalog number: 011656.30
also known as 011656-30
Price (USD)
Quantity
-

Chemical Identifiers

CAS
10294-39-0
IUPAC Name
barium(2+) trihydrate diperchlorate
Molecular Formula
BaCl2H6O11
InChI Key
NBHDMFAHVJWOHD-UHFFFAOYSA-L
SMILES
O.O.O.[Ba++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Assay (unspecified)
Ba 33.8-36.5%

Description

Barium perchlorate, trihydrate, reagent is an extremely powerful oxidizing agent, and therefore commonly used in industry and pyrotechnics. It is also used in RPE-For analysis. It is used as an oxidizing agent, for gas drying, for ribonuclease determination and in the water analysis for sulfate titration with Thorin as indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text