Sodium hexanitritocobaltate(III), ACS
Sodium hexanitritocobaltate(III), ACS
Sodium hexanitritocobaltate(III), ACS
Thermo Scientific Chemicals

Sodium hexanitritocobaltate(III), ACS

CAS: 14649-73-1 | CoN6Na3O12 | 403.933 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
產品號碼 011667.22
亦稱為 011667-22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS14649-73-1
IUPAC Namecobalt(3+) trisodium hexakis(dioxoamine)
Molecular FormulaCoH6N6Na3O12
InChI KeyQLNJFTGEDYRKNP-UHFFFAOYSA-N
SMILES[Na+].[Na+].[Na+].[Co+3].[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O
檢視更多
規格Specification Sheet規格表
Insoluble material0.02%
CommentSuitability for determination of potassium P.T.
Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only