Tungsten(VI) oxide, 99.8% (metals basis)
Tungsten(VI) oxide, 99.8% (metals basis)
Tungsten(VI) oxide, 99.8% (metals basis)
Thermo Scientific Chemicals

Tungsten(VI) oxide, 99.8% (metals basis)

CAS: 1314-35-8 | O3W | 231.837 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 011828.22
also known as 011828-22
Price (USD)
88.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
88.00
Each
Chemical Identifiers
CAS1314-35-8
IUPAC Nametrioxotungsten
Molecular FormulaO3W
InChI KeyZNOKGRXACCSDPY-UHFFFAOYSA-N
SMILESO=[W](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Total Metal Impurities≤0.2% (2000 ppm)
Particle Size10-25 Micron average
FormPowder
It is used in tungsten and tungstate manufacturing which are used as X-ray screens and for fire proofing fabrics. It is used as a ceramic pigment. Nanowires of Tungsten (VI) oxide are capable of absorbing a higher percentage of the sun radiation since it absorbs blue light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only