Thermo Scientific Chemicals

Manganese(II) oxide, Mn 76.0-78.0%, typically 99%, Thermo Scientific Chemicals

Catalog number: 011870.F7
5 x 2 kg, Each
Thermo Scientific Chemicals

Manganese(II) oxide, Mn 76.0-78.0%, typically 99%, Thermo Scientific Chemicals

Catalog number: 011870.F7
5 x 2 kg, Each
Quantity
Catalog number: 011870.F7
Price (USD)
Price: 463.00
Online price: 393.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1344-43-0
IUPAC Name
oxomanganese
Molecular Formula
MnO
InChI Key
VASIZKWUTCETSD-UHFFFAOYSA-N
SMILES
O=[Mn]
Assay (unspecified)
Mn 76.0-78.0%

Description

The main use of manganese dioxide is in dry-cell batteries, such as the alkaline battery and the zinc-carbon battery. It is useful as a pigment, and as a precursor to other manganese compounds. It is used as a reagent in organic synthesis, for instance, for the oxidation of allylic alcohols and arylmethyl alcohols. It is also employed in various industrial purposes such as in adhesives and sealant chemicals, lubricants and lubricant additives, fertilizers, oxidizing/reducing agents, plating agents and surface treating agents, processing aids, air care products, paints, and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text