Silver hexafluorophosphate, 98%
Silver hexafluorophosphate, 98%
Silver hexafluorophosphate, 98%
Thermo Scientific Chemicals

Silver hexafluorophosphate, 98%

CAS: 26042-63-7 | AgF6P | 252.832 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 011873.14
Price (EUR)
772,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
772,00
Each
Chemical Identifiers
CAS26042-63-7
IUPAC Namesilver(1+) hexafluoro-λ⁵-phosphanuide
Molecular FormulaAgF6P
InChI KeySCQBROMTFBBDHF-UHFFFAOYSA-N
SMILES[Ag+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
It is commonly used as an oxidant forming silver metal. It is often used as a reagent in inorganic and organometallic chemistry. It is used in the preparation of engineering novel materials as well as in preparation of supra molecular Ag(I) complexes. It is used in the development of new catalysts, replacing halide ligands with the weakly coordinating hexafluorophosphate anion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only