Thermo Scientific Chemicals

Silver iodide, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012111.14
25 g, Each
Thermo Scientific Chemicals

Silver iodide, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012111.14
25 g, Each
Quantity
Catalog number: 012111.14
also known as 012111-14
Price (USD)
Price: 204.00
Online price: 173.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7783-96-2
IUPAC Name
silver(1+) iodide
Molecular Formula
AgI
InChI Key
MSFPLIAKTHOCQP-UHFFFAOYSA-M
SMILES
[Ag+].[I-]
Total Metal Impurities
0.001% max.

Description

Silver iodide is used in photographic film. Used in medical devices as a sustained release antibacterial. It also used in as an antiseptic and in cloud seeding. It is used in proteomics research applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text