Thermo Scientific Chemicals

Silver iodide, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

產品號碼: 012111.22
100 g, Each
Thermo Scientific Chemicals

Silver iodide, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

產品號碼: 012111.22
100 g, Each
Quantity
產品號碼: 012111.22
亦稱為 012111-22
價格 (TWD)

化學識別

CAS
7783-96-2
IUPAC Name
silver(1+) iodide
Molecular Formula
AgI
InChI Key
MSFPLIAKTHOCQP-UHFFFAOYSA-M
SMILES
[Ag+].[I-]
Total Metal Impurities
0.001% max.

說明

Silver iodide is used in photographic film. Used in medical devices as a sustained release antibacterial. It also used in as an antiseptic and in cloud seeding. It is used in proteomics research applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text