Carburo de vanadio, 99 % (base metálica), Thermo Scientific Chemicals
Carburo de vanadio, 99 % (base metálica), Thermo Scientific Chemicals
Carburo de vanadio, 99 % (base metálica), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carburo de vanadio, 99 % (base metálica), Thermo Scientific Chemicals

CAS: 12070-10-9 | CV | 62.95 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Número de catálogo 012141.14
también denominado 012141-14
Precio (EUR)
69,40
Each
Cantidad:
25 g
Pedido a granel o personalizado
Precio (EUR)
69,40
Each
Identificadores químicos
CAS12070-10-9
IUPAC Namemethanidylidynevanadiumylium
Molecular FormulaCV
InChI KeyZLANVVMKMCTKMT-UHFFFAOYSA-N
SMILES[C-]#[V+]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Total Metal Impurities1% max.
Vanadium carbide is used as an additive to tungsten carbide to fine the carbide crystals to improve the property of the cermets. It used in cutting tools and the steel industry. Used as material for research in electronics. Used anode materials for lithium ion batteries, and in supercapacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only