Potassium pentachlororuthenate(III) hydrate, 99.95% (metals basis), Ru 25.4% min
Potassium pentachlororuthenate(III) hydrate, 99.95% (metals basis), Ru 25.4% min
Potassium pentachlororuthenate(III) hydrate, 99.95% (metals basis), Ru 25.4% min
Thermo Scientific Chemicals

Potassium pentachlororuthenate(III) hydrate, 99.95% (metals basis), Ru 25.4% min

CAS: 14404-33-2 | Cl5K2Ru | 356.52 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 012173.03
亦稱為 012173-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
規格
Assay Percent Notes(metals basis)
CAS14404-33-2
Chemical Name or MaterialPotassium pentachlororuthenate(III) hydrate
Concentration or Composition (by Analyte or Components)Ru 25.4% min.
Formula Weight356.54 (Anhydrous)
檢視更多
Potassium pentachlororuthenate(III) hydrate is used as a catalyst in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only