Thermo Scientific Chemicals

Barium acetate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Catalog number: 012198.A1
1 kg, Each
Thermo Scientific Chemicals

Barium acetate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Catalog number: 012198.A1
1 kg, Each
Quantity
Catalog number: 012198.A1
also known as 012198-A1
Price (USD)
Price: 150.00
Online price: 127.65
Your price:
Quantity
-

Chemical Identifiers

CAS
543-80-6
IUPAC Name
barium(2+) diacetate
Molecular Formula
C4H6BaO4
InChI Key
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES
[Ba++].CC([O-])=O.CC([O-])=O
Heavy Metals (as Pb)
≤5ppm
Insoluble material
≤ 0.01%
Assay (unspecified)
≥99.0 to ≤102.0% Ba(OOCCH₃)₂
Impurity content
Ca: ≤0.05%
Impurity content
Fe: ≤0.001%

Description

Barium acetate is used as a chemical reagent, textile mordant and lubricant. It is used to prepare other acetates and barium carbonate. It acts as a mordant for printing textile fabrics. It is utilized for drying paints and varnishes. It serves as a catalyst in organic synthesis. It plays an important role in the separation of glycosaminoglycans. Further, it acts as a cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. In addition, it is used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text