Thermo Scientific Chemicals

Strontium hydroxide octahydrate, Technical grade, Thermo Scientific Chemicals

Catalog number: 012199.A3
2 kg, Each
Thermo Scientific Chemicals

Strontium hydroxide octahydrate, Technical grade, Thermo Scientific Chemicals

Catalog number: 012199.A3
2 kg, Each
Quantity
Catalog number: 012199.A3
also known as 012199-A3
Price (USD)
Price: 125.00
Online price: 106.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1311-10-0
IUPAC Name
strontium(2+) octahydrate dihydroxide
Molecular Formula
H18O10Sr
InChI Key
UJPWWRPNIRRCPJ-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]
Assay (unspecified)
94% min

Description

Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text