Thermo Scientific Chemicals

Bismuth(III) nitrate pentahydrate, ACS, 98% min, Thermo Scientific Chemicals

Catalog number: 012231.A1
1 kg, Each
Thermo Scientific Chemicals

Bismuth(III) nitrate pentahydrate, ACS, 98% min, Thermo Scientific Chemicals

Catalog number: 012231.A1
1 kg, Each
Quantity
Catalog number: 012231.A1
also known as 012231-A1
Price (USD)
Quantity
-

Chemical Identifiers

CAS
10035-06-0
IUPAC Name
bismuth(3+) pentahydrate trinitrate
Molecular Formula
BiH10N3O14
InChI Key
FBXVOTBTGXARNA-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay (unspecified)
98.0% Bi(NO₃)₃?5H₂O min
Impurity content
Ag 0.001%
Impurity content
As 0.001%
Impurity content
Cl 0.001%
Impurity content
Cu 0.002%

Description

Luminous paints, preparation of bismuth saltsBismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text