Mercury(II) chloride, ACS, 99.5% min
Mercury(II) chloride, ACS, 99.5% min
Mercury(II) chloride, ACS, 99.5% min
Thermo Scientific Chemicals

Mercury(II) chloride, ACS, 99.5% min

CAS: 7487-94-7 | Cl2Hg | 271.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012274.14
also known as 012274-14
Price (EUR)
52,10
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
52,10
Each
Chemical Identifiers
CAS7487-94-7
IUPAC Namemercury(2+) dichloride
Molecular FormulaCl2Hg
InChI KeyLWJROJCJINYWOX-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentFe 0.002%
Assay (unspecified)99.5% Cl₂Hg min
CommentResidue after reduction 0.02%
CommentSolution in ethyl ether P.T.
Mercury(II) chloride is used as a catalyst in making plastics, depolarizer in batteries and as a reagent in organic synthesis and analytical chemistry. It is also used to prepare an amalgam with metals such as aluminum, which finds application as reducing agent in organic synthesis. It also finds application in photography as a photographic intensifier. Further, it serves as a preservative and used in the treatment of syphilis. It is also used to remove dithiane groups bonded to a carbonyl in an umpolung reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only