Thermo Scientific Chemicals

Barium fluoride, 99%, Thermo Scientific Chemicals

Catalog number: 012338.A1
1 kg, Each
Thermo Scientific Chemicals

Barium fluoride, 99%, Thermo Scientific Chemicals

Catalog number: 012338.A1
1 kg, Each
Quantity
Catalog number: 012338.A1
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7787-32-8
IUPAC Name
barium(2+) difluoride
Molecular Formula
BaF2
InChI Key
OYLGJCQECKOTOL-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ba++]
Appearance (Color)
Colorless or white
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥98.5%

Description

It finds applications in ceramic flux, glass manufacturing, metallurgy, electronics and dry-film lubricants. It is transparent from ultraviolet to infrared. Barium fluoride is usually employed as a scintillator material, and useful as a transmission window material for IR spectroscopy, mostly used for analysis of fuel oil samples. Of all the optical fluorides, it is the most resistant to high-energy radiation. It is used as a preopacifying agent, and in the production of enamel, welding agents, and glazing frits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text