Thermo Scientific Chemicals

Strontium zirconium oxide, 99.3% (metals basis), Thermo Scientific Chemicals

Catalog number: 012346.30
250 g, Each
Thermo Scientific Chemicals

Strontium zirconium oxide, 99.3% (metals basis), Thermo Scientific Chemicals

Catalog number: 012346.30
250 g, Each
Quantity
Catalog number: 012346.30
also known as 012346-30
Price (USD)

Chemical Identifiers

CAS
12036-39-4
IUPAC Name
strontium(2+) oxozirconiumbis(olate)
Molecular Formula
O3SrZr
InChI Key
FCCTVDGKMTZSPU-UHFFFAOYSA-N
SMILES
[Sr++].[O-][Zr]([O-])=O
Total Metal Impurities
0.7% max.

Description

Mostly as intermediates( primary and secondary ). For chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text