Barium titanium oxide, 99%
Barium titanium oxide, 99%
Barium titanium oxide, 99%
Thermo Scientific Chemicals

Barium titanium oxide, 99%

Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers | CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
50 g
500 g
2 kg
產品號碼 012348.A3
亦稱為 012348-A3
價格 (TWD)
-
Quantity:
2 kg
Request bulk or custom format
化學識別
CAS12047-27-7
IUPAC Namebarium(2+) oxotitaniumbis(olate)
Molecular FormulaBaO3Ti
InChI KeyWNKMTAQXMLAYHX-UHFFFAOYSA-N
SMILES[Ba++].[O-][Ti]([O-])=O
檢視更多
規格Specification Sheet規格表
Particle Size+325 Mesh Screen: ≤0.10%
Assay from Supplier's CofA(Ba+Sr)O: ≥65.0%
Composition(Ba+Sr)/Ti Ratio: 0.988-1.002
FormPowder
Assay from Supplier's CofATiO₂: ≥34.0%
Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only