Thermo Scientific™

Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific™

25 x 25 mm
Thermo Scientific™

Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific™

25 x 25 mm
Quantity
Catalog number: 012388.FF
also known as 012388-FF
Price (USD)
151.00 / -
Your Price:
Quantity
-

Chemical Identifiers

CAS
7440-52-0
IUPAC Name
erbium
Molecular Formula
Er
InChI Key
UYAHIZSMUZPPFV-UHFFFAOYSA-N
SMILES
[Er]
Total Rare Earth Oxide Impurities
0.1% max.

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Packaged under argon
RUO – Research Use Only

Figures