Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO)

CAS: 7440-54-2 | Gd | 157.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 012397.FI
also known as 012397-FI
Price (USD)
454.65
Special offer
Online exclusive
535.00
Save 80.35 (15%)
Each
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
454.65
Special offer
Online exclusive
535.00
Save 80.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7440-54-2
IUPAC Namegadolinium
Molecular FormulaGd
InChI KeyUIWYJDYFSGRHKR-UHFFFAOYSA-N
SMILES[Gd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only