Scandium foil, 0.25mm (0.01in) thick, 99.9% (REO)
Scandium foil, 0.25mm (0.01in) thick, 99.9% (REO)
Scandium foil, 0.25mm (0.01in) thick, 99.9% (REO)
Scandium foil, 0.25mm (0.01in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Scandium foil, 0.25mm (0.01in) thick, 99.9% (REO)

CAS: 7440-20-2 | Sc | 44.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 012424.FI
also known as 012424-FI
Price (EUR)
1.013,00
Each
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (EUR)
1.013,00
Each
Add to cart
Chemical Identifiers
CAS7440-20-2
IUPAC Namescandium
Molecular FormulaSc
InChI KeySIXSYDAISGFNSX-UHFFFAOYSA-N
SMILES[Sc]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only