Thermo Scientific™

Thulium pieces, sublimed dendritic, 99.9% (REO), Thermo Scientific™

5 g, Each
Thermo Scientific™

Thulium pieces, sublimed dendritic, 99.9% (REO), Thermo Scientific™

5 g, Each
Quantity
Catalog number: 012435.06
also known as 012435-06
Price (USD)
281.00 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
7440-30-4
Total Rare Earth Oxide Impurities
0.1% max.

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Packaged under argon
RUO – Research Use Only

Figures