Gallium Indium eutectic, 99.99% (metals basis)
Thermo Scientific Chemicals

Gallium Indium eutectic, 99.99% (metals basis)

Gallium Indium eutectic, Ga:In; 75.5:24.5 wt%, 10 g, 99.99% (metals basis), 6.2, Hazard Class: 8; Packaging Group: III, 231-163-8, H290-H314-H335, P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c, 15.7 deg.C, MFCD00144387, Liquid | Ga:In; 75.5:24.5 wt%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number 012478.03
also known as 012478-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Assay Percent Range99.99% (metals basis)
Chemical Name or MaterialGallium Indium eutectic
DOT InformationHazard Class: 8; Packaging Group: III
Density6.2
EINECS Number231-163-8
View more
It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices. Gallium-Indium eutectic top contact, and a halogen light source was used to test P3HT-PCBM organic solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only