Lead shot, 3mm (0.1in), 99.999% (metals basis)
Lead shot, 3mm (0.1in), 99.999% (metals basis)
Lead shot, 3mm (0.1in), 99.999% (metals basis)
Thermo Scientific Chemicals

Lead shot, 3mm (0.1in), 99.999% (metals basis)

CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 012511.18
also known as 012511-18
Price (EUR)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
FormShot, 3mm (0.1in).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only