Thermo Scientific Chemicals

Nickel(II) hydroxide, Ni 61%, Thermo Scientific Chemicals

Catalog number: 012517.A1
1 kg, Each
Thermo Scientific Chemicals

Nickel(II) hydroxide, Ni 61%, Thermo Scientific Chemicals

Catalog number: 012517.A1
1 kg, Each
Quantity
Catalog number: 012517.A1
also known as 012517-A1
Price (USD)
Price: 359.00
Online price: 305.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12054-48-7
IUPAC Name
nickel(2+) dihydroxide
Molecular Formula
H2NiO2
InChI Key
BFDHFSHZJLFAMC-UHFFFAOYSA-L
SMILES
[OH-].[OH-].[Ni++]
Assay (unspecified)
61% Ni min

Description

Due to its redox behavior, it finds use in rechargeable battery electrodes. It is useful for the storage of electrochemical energy, with potential applications in car batteries. It is a potential material for several biological applications, for example in chromosomal DNA quantification assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text