Cobalt(II) 2,4-pentanedionate
Cobalt(II) 2,4-pentanedionate
Cobalt(II) 2,4-pentanedionate
Thermo Scientific Chemicals

Cobalt(II) 2,4-pentanedionate

CAS: 14024-48-7 | C10H14CoO4 | 257.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 012537.30
also known as 012537-30
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS14024-48-7
IUPAC Nameλ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14CoO4
InChI KeyUTYYEGLZLFAFDI-FDGPNNRMSA-L
SMILES[Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple or pink
FormPowder or lumps or chunks
Insoluble material≤1%
Melting point155-165°C
Volatiles≤3.5%
Cobalt(II) 2,4-pentanedionate is used as an important catalyst for organic reactions. It is also used as paint dryer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only