Chromium(III) 2,4-pentanedionate, 97%
Chromium(III) 2,4-pentanedionate, 97%
Chromium(III) 2,4-pentanedionate, 97%
Thermo Scientific Chemicals

Chromium(III) 2,4-pentanedionate, 97%

Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD | CAS: 21679-31-2 | C15H21CrO6 | 349.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 012538.36
also known as 012538-36
Price (EUR)
174,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
174,00
Each
Chemical Identifiers
CAS21679-31-2
IUPAC Namechromium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21CrO6
InChI KeyJWORPXLMBPOPPU-LNTINUHCSA-K
SMILES[Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting PointReport Value
Insoluble material< 0.20%
Volatiles≤0.50%
FormPowder or crystals or crystalline powder
Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVDChromium(III) 2,4-pentanedionate is used in the reduction of detonation of nitromethane. It is also used in NMR spectroscopy as a relaxation agent due to its solubility in nonpolar organic solvents and its paramagnetism. Further, it is involved in the modification of the surface properties of solid polyurethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only