Citric acid, 99.5-100.5%, powder, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Citric acid, 99.5-100.5%, powder, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
수량:
2.5 kg
12 kg
카탈로그 번호 0127-05
제품 가격(KRW)
-
수량:
2.5 kg
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
CAS77-92-9
화학물질 이름 또는 재질Citric acid monohydrate
색상White
용융점139°C to 152°C
분자식C6H8O7·H2O
포장Poly bottle, tamper-evident cap
동의어Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
등급Multi-Compendial
분자량(g/mol)210.14
백분율 순도99.5 to 100.5%
수량2.5 kg
Unit SizeEach