Citric acid, 99.5-100.5%, powder, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Citric acid, 99.5-100.5%, powder, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
2.5 kg
12 kg
Número de catálogo 0127-07
Precio (MXN)
-
Cantidad:
12 kg
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
CAS77-92-9
Nombre del producto químico o materialCitric acid monohydrate
ColorBlanco
Punto de fusión139°C to 152°C
Fórmula molecularC6H8O7·H2O
EnvasePolyethylene pail, screw top
SinónimoHydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
GradoMulti-Compendial
Peso molecular (g/mol)210.14
Porcentaje de pureza99.5 to 100.5%
Cantidad12 kg
Unit SizeEach