Manganese(II) sulfide, 99.9% (metals basis)
Manganese(II) sulfide, 99.9% (metals basis)
Manganese(II) sulfide, 99.9% (metals basis)
Thermo Scientific Chemicals

Manganese(II) sulfide, 99.9% (metals basis)

CAS: 18820-29-6 | MnS | 87.00 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
產品號碼 012835.14
亦稱為 012835-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS18820-29-6
IUPAC Namemanganese(2+) sulfanediide
Molecular FormulaMnS
InChI KeyVCTOKJRTAUILIH-UHFFFAOYSA-N
SMILES[S--].[Mn++]
檢視更多
規格Specification Sheet規格表
Total Metal Impurities0.1% max.

it is used as an intermediate in chemical research& pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only