Lithium sulfide, 99.9% (metals basis)
Lithium sulfide, 99.9% (metals basis)
Lithium sulfide, 99.9% (metals basis)
Lithium sulfide, 99.9% (metals basis)
Thermo Scientific Chemicals

Lithium sulfide, 99.9% (metals basis)

CAS: 12136-58-2 | Li2S | 45.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
250 g
Catalog number 012839.04
also known as 012839-04
Price (EUR)
55,60
Each
Quantity:
2 g
Request bulk or custom format
Price (EUR)
55,60
Each
Chemical Identifiers
CAS12136-58-2
IUPAC Namedilithium(1+) sulfanediide
Molecular FormulaLi2S
InChI KeyGLNWILHOFOBOFD-UHFFFAOYSA-N
SMILES[Li+].[Li+].[S--]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormPowder
Total Metal Impurities≤0.1%
It act as a superconductor and widely used in lithium-sulfur batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

How should Lithium sulfide, 99.9% (metals basis) (Cat. No. 012839.xx) be stored and handled?

Lithium sulfide releases flammable gas when in contact with water and should be kept away from water, moist air, or acids. Lithium sulfide should be stored and handled under a dry, inert gas atmosphere. Store at ambient temperatures, typically 20-25 degrees C.