Thermo Scientific Chemicals

Barium sulfide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 012848.18
50 g, Each
Thermo Scientific Chemicals

Barium sulfide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 012848.18
50 g, Each
Quantity
Catalog number: 012848.18
also known as 012848-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
21109-95-5
IUPAC Name
barium(2+) sulfanediide
Molecular Formula
BaS
InChI Key
CJDPJFRMHVXWPT-UHFFFAOYSA-N
SMILES
[S--].[Ba++]
Total Metal Impurities
0.3% max.

Description

Barium sulfide has different characters, so it is used in a variety of ways, such as in electronics, paint pigments, dehairing hides, flame retardant, luminous paints, and generating pure hydrogen sulfide. Barium sulfide can deviate X-rays and helps to give clearly image of the soft tissue. Barium sulfide is a precursor to other barium compounds and also used as a short wavelength emitters for electronic displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text