Samarium powder, -40 mesh, 99.9% (REO)
Samarium powder, -40 mesh, 99.9% (REO)
Samarium powder, -40 mesh, 99.9% (REO)
Samarium powder, -40 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Samarium powder, -40 mesh, 99.9% (REO)

CAS: 7440-19-9 | Sm | 150.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
10 g
Catalog number 012895.18
also known as 012895-18
Price (USD)/ Each
606.65
Special offer
Online exclusive
714.00
Save 107.35 (15%)
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
606.65
Special offer
Online exclusive
714.00
Save 107.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Namesamarium
Molecular FormulaSm
InChI KeyKZUNJOHGWZRPMI-UHFFFAOYSA-N
SMILES[Sm]
Molecular Weight (g/mol)150.36
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only