Erbium(III) nitrate hydrate, 99.9% (REO)
Erbium(III) nitrate hydrate, 99.9% (REO)
Erbium(III) nitrate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Erbium(III) nitrate hydrate, 99.9% (REO)

CAS: 10031-51-3 | ErH10N3O14 | 443.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 012920.14
also known as 012920-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10031-51-3
IUPAC Nameerbium(3+) pentahydrate trinitrate
Molecular FormulaErH10N3O14
InChI KeyLWHHUEHWVBVASY-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Used as laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only