Yttrium Cobalt lump, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Yttrium Cobalt lump, REacton™, 99.9% (REO)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 012952.14
also known as 012952-14
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Specifications
Assay Percent Notes(REO)
Avg. Mol. Wt. or Mol. Wt. RangeY:Co; 84:15 wt% (Ca 1%)
Chemical Name or MaterialYttrium Cobalt lump
Health Hazard 1H317-H334
Health Hazard 2GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only