Thermo Scientific Chemicals

Calcium acetate hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

Catalog number: 012974.14
25 g, Each
Thermo Scientific Chemicals

Calcium acetate hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

Catalog number: 012974.14
25 g, Each
Quantity
Catalog number: 012974.14
also known as 012974-14
Price (USD)
Price: 274.00
Online price: 236.65
Your price:
Quantity
-

Chemical Identifiers

CAS
114460-21-8
IUPAC Name
calcium diacetate
Molecular Formula
C4H6CaO4
InChI Key
VSGNNIFQASZAOI-UHFFFAOYSA-L
SMILES
[Ca++].CC([O-])=O.CC([O-])=O
Total Metal Impurities
0.003% max.

Description

Calcium acetate has been utilized in the crystallization of proteins, such as human annexin II and the bacterial heme oxygenase (Hmu O), and of a complex of human CDK6 with a virus-encoded cyclin. It has been used in the manufactureof metallic soaps and synthetic resins, in lubricants as a corrosion inhibitor, and as a mordant in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text