Dodecacarbonyltetracobalt, 98% min, Thermo Scientific Chemicals
Dodecacarbonyltetracobalt, 98% min, Thermo Scientific Chemicals
Dodecacarbonyltetracobalt, 98% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dodecacarbonyltetracobalt, 98% min, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
013055.06
also known as 013055-06
5 g
Catalog number 013055.06
also known as 013055-06
Price (USD)
383.65
Online Exclusive
426.00
Save 42.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
383.65
Online Exclusive
426.00
Save 42.35 (10%)
Each
Chemical Identifiers
CAS17786-31-1
IUPAC Namedodecakis(methanidylidyneoxidanium) tetracobalt
Molecular FormulaC12Co4O12
InChI KeyYMFAWOSEDSLYSZ-UHFFFAOYSA-N
SMILES[Co].[Co].[Co].[Co].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only