Hexacarbonylchromium, 99%
Hexacarbonylchromium, 99%
Hexacarbonylchromium, 99%
Thermo Scientific Chemicals

Hexacarbonylchromium, 99%

CAS: 13007-92-6 | C6CrO6 | 220.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number 013061.03
also known as 013061-03
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.20
Save 7.55 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.20
Save 7.55 (14%)
Each
Chemical Identifiers
CAS13007-92-6
IUPAC Namehexakis(methanidylidyneoxidanium) chromium
Molecular FormulaC6CrO6
InChI KeyKOTQLLUQLXWWDK-UHFFFAOYSA-N
SMILES[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Hexacarbonylchromium is widely used as a catalyst for isomerization of alkenes. It is involved in the hydrogenation of 1,3-conjugated dienes and oxidation of allylic and benzylic positions. It acts as a precursor to prepare phenanthrenes, tricarbonyl(arene)chromium and Fischer carbine complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only