Thermo Scientific Chemicals

Sodium hexafluorophosphate, 99+%, Thermo Scientific Chemicals

Catalog number: 013064.22
100 g, Each
Thermo Scientific Chemicals

Sodium hexafluorophosphate, 99+%, Thermo Scientific Chemicals

Catalog number: 013064.22
100 g, Each
Quantity
Catalog number: 013064.22
Price (USD)
Price: 369.00
Online price: 313.65
Your price:
Quantity
-

Chemical Identifiers

CAS
21324-39-0
IUPAC Name
sodium hexafluoro-λ⁵-phosphanuide
Molecular Formula
F6NaP
InChI Key
KMADQUOCJBLXRP-UHFFFAOYSA-N
SMILES
[Na+].F[P-](F)(F)(F)(F)F
Assay (unspecified)
99.1% min

Description

Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text