Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

CAS: 181266-82-0 | C15H21EuO6 | 449.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
5 g
10 g
Catalog number 013209.04
also known as 013209-04
Price (USD)
142.65
Special offer
Online exclusive
Ends: 30-Jun-2026
168.00
Save 25.35 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
142.65
Special offer
Online exclusive
Ends: 30-Jun-2026
168.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS181266-82-0
IUPAC Nameeuropium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21EuO6
InChI KeyYMTKZYSHNNXTIJ-UHFFFAOYSA-N
SMILES[Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only