Molybdenum pellet, 99.7% (metals basis)
Molybdenum pellet, 99.7% (metals basis)
Molybdenum pellet, 99.7% (metals basis)
Molybdenum pellet, 99.7% (metals basis)
Thermo Scientific Chemicals

Molybdenum pellet, 99.7% (metals basis)

Approx. 12.5 x 18 x 25mm, HotPressed, density Approx. 8.0g/cm3 | CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number 013242.A3
also known as 013242-A3
Price (USD)/ Each
475.65
Online Exclusive
528.00
Save 52.35 (10%)
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
475.65
Online Exclusive
528.00
Save 52.35 (10%)
Add to cart
Chemical Identifiers
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
Molecular Weight (g/mol)95.95
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only